Learn More
CD25 Mouse anti-Human, PE, Clone: M-A251, BD
Mouse Monoclonal Antibody
€ 67.45 - € 280.00
Specifications
| Antigen | CD25 |
|---|---|
| Clone | M-A251 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15800669
|
BD Biosciences
560989 |
25 Tests |
€ 67.50 € 67.45 / Each Save € 0.05 0.07% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15888158
|
BD Biosciences
555432 |
100 Tests |
€ 280.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The M-A251 monoclonal antibody specifically binds to the 55 kDa type I transmembrane glycoprotein known as the low-affinity interleukin-2 receptor alpha chain subunit (IL-2Rα). CD25 is expressed on regulatory T cells and on activated lymphocytes (T and B) and monocytes. It associates with the IL-2Rβ/CD122 and the IL-2Rγ/CD132 receptor chains to form the high-affinity IL-2R complex. CD25 expression on T and B lymphocytes is upregulated by antigenic or mitogenic stimulation. Soluble CD25/IL-2Rα is produced as a consequence of lymphocyte stimulation and is found in biological fluids following inflammatory responses.
Specifications
| CD25 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| M-A251 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.