Learn More
Invitrogen™ CD252 (OX40 Ligand) Monoclonal Antibody (RM134L), Alexa Fluor™ 488, eBioscience™

Rat Monoclonal Antibody
€ 395.00
Specifications
| Antigen | CD252 (OX40 Ligand) |
|---|---|
| Clone | RM134L |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: The RM134L monoclonal antibody reacts with mouse CD252 also known as OX-40 Ligand, a member of the TNF superfamily. OX-40L is induced on mouse splenic B cells stimulated with a combination of anti-IgM and anti-CD40. Neither resting nor activated mouse T cells express OX-40L. The interaction of OX-40 (CD134) with OX-40L has been implicated in T cell-dependent humoral response. RM134L inhibits the costimulatory activity of mouse OX-40L transfected cells for anti-CD3-stimulated T cell proliferation. Applications Reported: This RM134L antibody has been reported for use in flow cytometric analysis. Applications Tested: This RM134L antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.
The tumor necrosis factor superfamily member TNFSF4 is a type II membrane bound, non-covalently linked homotrimeric protein. It is expressed on antigen presenting cells, such as dendritic cells and activated B-cells, and also on other cells such as vascular endothelial cells, mast cells, and natural killer cells. TNFSF4 signals specifically through the TNFRSF4 receptor, is expressed predominantly on CD4+T cells but also on certain activated CD8+T cells. TNFRSF4/TNFSF4 functions as a costimulatory signal, which is required for a productive interaction between antigen presenting cells and their target T-cells. It enhances cell proliferation and survival, and increases expression of RANTES, IL-2, IL-3, and IFNgamma. TNFRSF4/TNFSF4 signaling plays an important role in immunotolerance.Specifications
| CD252 (OX40 Ligand) | |
| 0.5 mg/mL | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Rat | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P43488 | |
| 22164 | |
| Primary | |
| Affinity chromatography |
| RM134L | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Mouse | |
| Tnfsf4 | |
| Ath1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; glycoprotein Gp34; GP34; OX40 antigen ligand; OX40 ligand; OX40L; OX-40L; OX4OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; TNFSF4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; TXGP1; Txgp1l | |
| Tnfsf4 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.