Learn More
Invitrogen™ CD268 (BAFF Receptor) Monoclonal Antibody (2C4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528263
Description
This antibody recognizes the chicken homologue of human CD184, also known as CXCR4 (C-X-C Receptor type 4) and fusin. CXCR4 is a receptor for the chemokine, stromal-derived factor-1 (SDF-1), also known as CXCL12. Bursa cells may be used as a positive control for CXCR4 in flow cytometry.
Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), also known as BAFFR, is a protein in humans is encoded by the TNFRSF13C gene. The BAFFR gene is mapped to chromosome 22q13. 1-q13. 31. It has got 184 amino acid transmembrane protein which is 56% identical to the mouse protein. B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. BAFF plays a crucial role in B cell development and can function through receptors other than BCMA.
Specifications
| CD268 (BAFF Receptor) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with <0.1% sodium azide | |
| 0 | |
| TNFRSF13C | |
| Chicken BAFF-R transfected HEK293 cells. | |
| 100 μg | |
| Primary | |
| Chicken | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 2C4 | |
| Unconjugated | |
| TNFRSF13C | |
| 2010006P15Rik; B cell-activating factor receptor; BAFF receptor; BAFFR; BAFF-R; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; Bcmd; Bcmd1; Bcmd-1; BLyS receptor 3; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNF receptor superfamily member 13C; Tnfrsf13c; Tumor necrosis factor receptor superfamily member 13C; tumor necrosis factor receptor superfamily, member 13C | |
| Mouse | |
| Protein G | |
| RUO | |
| 417983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.