missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Ligand/TNFSF7/CD70 Antibody (cusatuzumab), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28627CL1
This item is not returnable.
View return policy
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal antibody specifically detects CD27 Ligand/TNFSF7/CD70 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD27 Ligand/TNFSF7/CD70 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
| TNFSF7/ CD27L/ CD70 | |
| 0.1 mL | |
| Apoptosis | |
| 970 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| cusatuzumab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction