missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Ligand/TNFSF7/CD70 Antibody (TAN 1-7), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50150AF750
This item is not returnable.
View return policy
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal antibody specifically detects CD27 Ligand/TNFSF7/CD70 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Functional Assay, CyTOF
Specifications
| CD27 Ligand/TNFSF7/CD70 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
| Recombinant mouse CD27 Ligand/TNFSF7/CD70 (extracellular domain, aa residues 41-195) | |
| 0.1 mL | |
| Apoptosis | |
| 970 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, ELISA, Immunofluorescence, Functional Assay, CyTOF | |
| TAN 1-7 | |
| 50 mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction