missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Ligand/TNFSF7/CD70 Antibody (TNFS7/1026), Novus Biologicals™
Mouse Monoclonal Antibody
€ 277.00 - € 474.00
Specifications
| Antigen | CD27 Ligand/TNFSF7/CD70 |
|---|---|
| Clone | TNFS7/1026 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunoprecipitation 0.5 - 1 ug/500ug Protein Lysate, Immunohistochemistry-Frozen 0.5 - 1 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18159498
|
Novus Biologicals
NBP2-44546-0.02MG |
0.02 mg |
€ 277.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18168727
|
Novus Biologicals
NBP2-44546-0.1MG |
0.1 mg |
€ 474.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunofluorescence
CD27 Ligand/TNFSF7/CD70 Monoclonal specifically detects CD27 Ligand/TNFSF7/CD70 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Functional, Immunofluorescence.Specifications
| CD27 Ligand/TNFSF7/CD70 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 970 | |
| Recombinant human CD70 protein | |
| Primary | |
| Store at 4C. | |
| 30 kDa |
| TNFS7/1026 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunoprecipitation 0.5 - 1 ug/500ug Protein Lysate, Immunohistochemistry-Frozen 0.5 - 1 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
| CD70 | |
| IgG1 κ | |
| Protein A or G purified | |
| It recognizes a protein of 30kDa, identified as CD70. It is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T- and B-lymphocytes. It induces proliferation of co-stimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title