Learn More
CD27 Mouse, anti-Human, Unconjugated, Clone: O323, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65132-1-IG-500UG
Description
CD27 (also known as TNFRSF7) is a type I glycoprotein expressed on some B cells and the majority of T cells, and is a member of the tumor necrosis factor (TNF) receptor family. CD27 is required for generation and long-term maintenance of T cell immunity (PMID: 11062504). It is a receptor for CD70 (CD27L). Ligation of CD27 by CD70 induces strong ubiquitination of TRAF and the activation of both canonical and non-canonical NF-kappaB pathways, as well as the JNK pathway (PMID: 19426224). CD27 may also play a role in apoptosis through association with SIVA1.
Specifications
| CD27 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| O323 | |
| Unconjugated | |
| 100 μg | |
| 939 | |
| Liquid |