Learn More
CD27/TNFRSF7 Antibody (LG.7F9), DyLight 488, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand:  Novus Biologicals NBP1-43428G
Product Code. 18288972
  • ONLINE EXCLUSIVE PRICE
    € 554.00
    € 523.95 / 0.10mL
    Save € 30.05
    5% Off
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD27/TNFRSF7
Monoclonal
DyLight 488
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Armenian Hamster
Affinity Purified
RUO
Primary
Human, Mouse, Rat
IgG
Flow Cytometry
LG.7F9
Flow Cytometry
CD27
The immunogen for this antibody was CD27.
0.1 mL
Apoptosis, Cancer, Immunology, Innate Immunity
939
Store at 4C in the dark.