missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27/TNFRSF7 Antibody (LPFS2/8838R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23582-20ug
This item is not returnable.
View return policy
Description
CD27/TNFRSF7 Monoclonal antibody specifically detects CD27/TNFRSF7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD27/TNFRSF7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
| Recombinant human full-length CD27/TNFRSF7 protein (exact sequence is proprietary) | |
| 20 μg | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cells | |
| 939 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| LPFS2/8838R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction