Learn More
Novus Biologicals™ PAFAH1B3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-29675
Product Code. 30044224
  • € 571.00 / 100µg
Description
PAFAH1B3 Polyclonal antibody specifically detects PAFAH1B3 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PAFAH1B3
Polyclonal
Tris-citrate/phosphate pH 7 -8.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Immunoprecipitation, Western Blot
Unconjugated
EC 3.1.1.47, FLJ44990, PAF acetylhydrolase 29 kDa subunit, PAF-AH 29 kDa subunit, PAFAH subunit gamma, PAF-AH subunit gamma, PAF-AH1b alpha 1 subunit, PAFAHG, platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa), platelet-activating factor acetylhydrolase IB subunit gamma, platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit (29kD), platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa)
Between 1 and 50
100 μg
Neuroscience
5050
Store at 4°C. Do not freeze.