missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27/TNFRSF7 Antibody (rLPFS2/8836), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24087MFV610
This item is not returnable.
View return policy
Description
CD27/TNFRSF7 Monoclonal antibody specifically detects CD27/TNFRSF7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD27/TNFRSF7 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
| Recombinant human CD27/TNFRSF7 protein fragment (aa 28-170) (exact sequence is proprietary) | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cells | |
| 939 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rLPFS2/8836 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction