missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27/TNFRSF7 Antibody (rLPFS2/8836), Novus Biologicals™
Mouse Monoclonal Antibody
€ 245.00 - € 486.00
Specifications
| Antigen | CD27/TNFRSF7 |
|---|---|
| Clone | rLPFS2/8836 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677804
|
Novus Biologicals
NBP3-23942-100ug |
100 μg |
€ 486.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645334
|
Novus Biologicals
NBP3-23942-20ug |
20 μg |
€ 245.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD27/TNFRSF7 Monoclonal antibody specifically detects CD27/TNFRSF7 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD27/TNFRSF7 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cells | |
| 10 mM PBS with 0.05% BSA | |
| 939 | |
| IgG1 κ | |
| Protein A or G purified |
| rLPFS2/8836 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
| Recombinant human CD27/TNFRSF7 protein fragment (aa 28-170) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title