missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD275 (B7-H2) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122121
This item is not returnable.
View return policy
Description
CD275 (B7-H2) Polyclonal Antibody for Western Blot, IHC (P)
Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.
Specifications
| CD275 (B7-H2) | |
| Polyclonal | |
| Unconjugated | |
| ICOSLG | |
| AU044799; B7 homolog 2; B7 homologue 2; B7h; B7H2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP1; B7RP-1; BG071784; CD275; GI50; GL50; GL50-B; ICOS ligand; ICOSL; ICOS-L; ICOSLG; inducible T-cell costimulator ligand; inducible T-cell co-stimulator ligand; KIAA0653; LICOS; Ly115l; mKIAA0653; RGD1562791; transmembrane protein B7-H2 ICOS ligand | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23308 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O75144 | |
| ICOSLG | |
| Recombinant protein Inducible T-Cell Co Stimulator Ligand. The antigen corresponds to amino acid range Ala1655-Asp1918 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction