Learn More
CD279 Mouse anti-Human, FITC, Clone: EH12.2H7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech FITC-65168-100TESTS
Description
The CD279 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD279. This antibody recognizes non-human primates, human antigen. The CD279 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
| CD279 | |
| Monoclonal | |
| 4 μL/Test | |
| Mouse | |
| Primary | |
| Human, Non-human Primate |
| Flow Cytometry | |
| EH12.2H7 | |
| FITC | |
| 100 Tests | |
| 5133 | |
| IgG1 κ |