Learn More
CD279 Mouse anti-Human, FITC, Clone: EH12.2H7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech FITC-65168-100TESTS
Product Code. 16821236
  • € 118.00 / 100 tests
Description
The CD279 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD279. This antibody recognizes non-human primates, human antigen. The CD279 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
Specifications
CD279
Monoclonal
4 μL/Test
Mouse
Primary
Human, Non-human Primate
Flow Cytometry
EH12.2H7
FITC
100 Tests
5133