missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 (PD-1) Rat anti-Mouse, BB700, Clone: RMP1-30, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 748242
This item is not returnable.
View return policy
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD279 (PD-1) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| CD279 (PD-1) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| RMP1-30 | |
| Brilliant Blue 700 (BB700) | |
| 50 μg | |
| Primary | |
| Mouse | |
| IgG2b κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?