missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 (PD-1) Rat anti-Mouse , PE-CF594, Clone: RMP1-30, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 566833
This item is not returnable.
View return policy
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD279 (PD-1) | |
| Monoclonal | |
| PE-CF594 | |
| PD-1; mPD-1; Pdc1; Pdcd1; Ly101 | |
| Mouse PD-1 Recombinant Protein | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| RMP1-30 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction