missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93558JF646
This item is not returnable.
View return policy
Description
CD28 Polyclonal antibody specifically detects CD28 in Human samples. It is validated for Western Blot
Specifications
| CD28 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| Synthetic peptide corresponding to residues 72-85 [SQQLQVYSKTGFN], which is the Ig-V-like region of human CD28. | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 940 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction