missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Antibody (JJ316), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81052MFV450
This item is not returnable.
View return policy
Description
CD28 Monoclonal antibody specifically detects CD28 in Rat samples. It is validated for Flow Cytometry
Specifications
| CD28 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Flow Cytometry | |
| JJ316 | |
| Flow Cytometry | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| Made by immunizing BALB/c mice with rat CD28 cDNA-transfected A20/J cells. | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 940 | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction