missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Antibody (YTH913.12), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64867B
This item is not returnable.
View return policy
Description
CD28 Monoclonal specifically detects CD28 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD28 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CD28 | |
| Human peripheral blood T-cells | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 940 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Frozen) | |
| YTH913.12 | |
| Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64867 recognizes the human CD28 cell surface antigen, a 44kD glycoprotein expressed by a subset of T lymphocytes. The antibody enhances the MLR response. It has been reported that this clone also recognizes an epitope of CD28 expressed by NK cells, which is not recognized by clones of 9.3 or CD28.2.(2) | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction