missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cd28, Hamster anti-Mouse, PerCP-Cy5.5, Clone: 37.51, Abnova™
Hamster Monoclonal Antibody
Brand: Abnova MAB15438.100ug
This item is not returnable.
View return policy
Specifications
| Cd28 | |
| Monoclonal | |
| PerCP-Cyanine5.5 | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| - | |
| Hamster | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2 |
| Flow Cytometry | |
| 37.51 | |
| Flow Cytometry The optimal working dilution should be determined by the end user. | |
| CD28 antigen | |
| Cd28 | |
| Mouse EL-4 (T-cell lymphoma) cells. | |
| RUO | |
| 12487 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction