Learn More
CD28 Monoclonal Antibody (37.51), Biotin, eBioscience™, Invitrogen™
Syrian Hamster Monoclonal Antibody
€ 198.00 - € 282.00
Specifications
Antigen | CD28 |
---|---|
Clone | 37.51 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15219519
|
Affymetrix eBioscience
13-0281-82 |
100 μg |
€ 198.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15229519
|
Affymetrix eBioscience
13-0281-85 |
500 μg |
€ 282.00
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 37.51 monoclonal antibody reacts with the mouse CD28 molecule, a 45 kDa homodimer expressed by thymocytes, mature T cells and NK cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent costimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells. Applications Reported: The 37.51 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 37.51 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Note: It has been observed that increased incubations times (30-45 minutes) with the CD28 antibody can enhance the staining obtained. Filtration: 0.2 μm post-manufacturing filtered.
CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary's Disease.Specifications
CD28 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P31041 | |
Cd28 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cd28 |
37.51 | |
Flow Cytometry | |
Biotin | |
Syrian Hamster | |
Mouse | |
12487 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.