missing translation for 'onlineSavingsMsg'
Learn More

CD28 Mouse anti-Human, Clone: C28/1636, Abnova™

Product Code. 16043016
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16043016 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16043016 Supplier Abnova Supplier No. MAB14703.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human CD28.

  • High specificity
  • High reactivity

Specifications

Antigen CD28
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone C28/1636
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human CD28.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA and 0.05% azide).
Gene Alias MGC138290/Tp44
Gene Symbols CD28
Host Species Mouse
Immunogen Recombinant protein corresponding to full-length human CD28.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 940
Test Specificity Recognizes a glycoprotein of 44-88kDa which is identified as CD28.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.