Learn More
CD28 Mouse Anti-Rat, Brilliant Violet 711, Clone: JJ319, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742586
Description
The JJ319 monoclonal antibody specifically recognizes CD28, which is expressed on virtually all T lymphocytes bearingαβT-cell receptors (TCR), on mostγδTCR-bearing T cells, and on a subset of NK cells. In the thymus, CD28 expression is developmentally regulated during the maturation ofαβTCR-bearing T cells. CD28 is a costimulatory receptor required for activation of T cells; its ligands include CD80 (B7-1) and CD86 (B7-2). Soluble JJ319 mAb costimulates the proliferative responses and IL-2 production of CD4+ and CD8+ T cells activated by anti-αβ-TCR mAb R73 (Cat. no. 554910). The alternate anti-rat CD28 mAb JJ316 (Cat. No. 554992) is capable of directly stimulating T cells in vitro and in vivo.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications
| CD28 | |
| JJ319 | |
| Brilliant Violet 711 | |
| Cd28, CD28RNA | |
| Rat CD28-transfected cell line | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.