missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Polyclonal Antibody

CD28 Polyclonal Antibody
Brand: Thermo Scientific PA5114522
This item is not returnable.
View return policy
Description
CD28 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| CD28 | |
| Polyclonal | |
| Unconjugated | |
| Cd28 | |
| antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 940 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P10747 | |
| Cd28 | |
| CD28 antibody was raised against a peptide corresponding to 18 amino acids near the carboxy terminus of human CD28. The immunogen is located within the last 50 amino acids of CD28. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction