missing translation for 'onlineSavingsMsg'
Learn More

CD29 (Integrin beta 1) Monoclonal Antibody (eBioHMb1-1 (HMb1-1)), APC-eFluor™ 780, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody

€ 348.00

Specifications

Antigen CD29 (Integrin beta 1)
Clone eBioHMb1-1 (HMb1-1)
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15341620
View Documents Promotion Details
Affymetrix eBioscience
47-0291-82
100 μg
€ 348.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 09-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The eBioHMb1-1 monoclonal antibody reacts with mouse and rat CD29 (integrin beta 1), a 110-120 kDa member of the beta integrin family expressed by leukocytes, endothelial, smooth muscle and epithelial cells. CD29 binds non-covalently with the alpha integrins CD49a-f to form the VLA-1 through VLA-6 complexes, as well as with CD51. These alpha-beta integrin heterodimers are capable of mediating a variety of cellular responses including adhesion, trafficking, proliferaton and differentiation. All integrins which include CD29 bind to extracellular matrix proteins including collagen, laminin, fibronectin and vitronectin, whereas some CD29-containing integrins can also interact with cellular receptors such as VCAM-1 and MadCAM-1. Applications Reported: This eBioHMb1-1 (HMb1-1) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioHMb1-1 (HMb1-1) antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633 nm).

CD29 (Integrin beta 1) Monoclonal antibody specifically detects CD29 (Integrin beta 1) in Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD29 (Integrin beta 1)
0.2 mg/mL
Monoclonal
Liquid
RUO
P09055, P49134
IgG
Affinity chromatography
Antibody
eBioHMb1-1 (HMb1-1)
Flow Cytometry
APC-eFluor 780
Armenian Hamster
Mouse, Rat
16412, 24511
Primary
4° C, store in dark, DO NOT FREEZE!
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.