missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD298 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 159.00 - € 448.00
Specifications
| Antigen | CD298 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17700834
![]() |
Invitrogen™
PA5119425 |
200 μL |
€ 448.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17773514
|
Invitrogen™
PA5122119 |
20 μL |
€ 159.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD298 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 3 subunit. This gene encodes a beta 3 subunit. A pseudogene exists for this gene, and it is located on chromosome 2.Specifications
| CD298 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| ATP1B3 | |
| AA409958; AI664000; Atp1b3; ATPase Na+/K+ transporting subunit beta 3; ATPase, Na+/K+ beta 3 polypeptide; ATPase, Na+/K+ transporting, beta 3 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 3; ATPB-3; AW212096; CD298; CD298 antigen; Na K-ATPase beta-3 subunit; Na+/K+ -ATPase beta 3 subunit; NKAB3S; Sodium Potassium ATPase; Sodium/potassium-dependent ATPase subunit beta-3; sodium/potassium-transporting ATPase subunit beta-3 | |
| ATP1B3 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P54709 | |
| 483 | |
| Recombinant protein Sodium/potassium Transporting ATPase Subunit Beta-3. The antigen corresponds to amino acid range 61-273 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title