Learn More
Invitrogen™ CD2AP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596807
Description
Positive Samples: A-431, Raji, MCF7, SW480, HL-60, Mouse kidney, Rat kidney; Cellular Location: Cell projection, Cytoplasm, cytoskeleton, ruffle Immunogen sequence: KYFSLKPEEK DEKSTLEQKP SKPAAPQVPP KKPTPPTKAS NLLRSSGTVY PKRPEKPVPP PPPIAKINGE VSSISSKFET EPVSKLKLDS EQLPLRPKSV DFDSLTVRTS KETDVVNFDD IASSENLLHL TANRPKMPGR RLPGRFNGGH SPTHSPEKIL KLPKEEDSAN LKPSELKKDT CYSPKPSVYL STPSSASKAN TTAFLTPLEI KAKVETDDVK KNSLDELRAQ IIELLCIVEA LKKDHGKELE KLRKDLEEEK TMRSNLEMEI EKLKKAVLSS.
This gene encodes a scaffolding molecule that regulates the actin cytoskeleton. The protein directly interacts with filamentous actin and a variety of cell membrane proteins through multiple actin binding sites, SH3 domains, and a proline-rich region containing binding sites for SH3 domains. The cytoplasmic protein localizes to membrane ruffles, lipid rafts, and the leading edges of cells. It is implicated in dynamic actin remodeling and membrane trafficking that occurs during receptor endocytosis and cytokinesis. Haploinsufficiency of this gene is implicated in susceptibility to glomerular disease.
Specifications
| CD2AP | |
| Polyclonal | |
| Unconjugated | |
| CD2AP | |
| adapter protein CMS; AL024079; C78928; Cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2 associated protein; Cd2ap; CD2-associated protein; CMS; mesenchyme to epithelium transition protein, SH3 domains; mesenchyme-to-epithelium transition protein with SH3 domains 1; Mets1; METS-1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12488, 23607, 316258 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| F1LRS8, Q9JLQ0, Q9Y5K6 | |
| CD2AP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 360-639 of human CD2AP (NP_0362521). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |