missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2F-10/SLAMF9 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76534H
This item is not returnable.
View return policy
Description
CD2F-10/SLAMF9 Polyclonal antibody specifically detects CD2F-10/SLAMF9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CD2F-10/SLAMF9 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| CD2 family member 10, CD2F10, CD2F-10MGC88194, CD84 homolog 1, CD84H1, CD84-H1cluster of differentiation 2 antigen family member 10, cluster of differentiation 84 homolog 1, SF2001, signaling lymphocytic activation molecule family member 2001, signaling lymphocytic activation molecule family member 9, SLAM family member 9 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human SLAMF9. The immunogen is located within the last 50 amino acids of SLAMF9. Amino Acid Squence: RMKKLMRNRMKLRKEAKPG | |
| 0.1 mL | |
| Immunology | |
| 89886 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction