missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 epsilon Antibody (145-2C11), HRP - Chimeric, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52693H
This item is not returnable.
View return policy
Description
CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, CyTOF
Specifications
| CD3 epsilon | |
| Monoclonal | |
| HRP | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| H-2Kb-specific murine cytotoxic T-lymphocyte (CTL) clone. | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 916 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, CyTOF | |
| 145-2C11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction