missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 epsilon Antibody (APA1/1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
€ 542.00
Specifications
| Antigen | CD3 epsilon |
|---|---|
| Clone | APA1/1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Biotin |
Description
CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOFSpecifications
| CD3 epsilon | |
| Western Blot | |
| Biotin | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| PBS | |
| 916 | |
| IgG1 | |
| Protein A purified |
| APA1/1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Purified human CD3 proteins isolated from thymus | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title