missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 epsilon Antibody (TR66), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97446PCP
This item is not returnable.
View return policy
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry.
Specifications
| CD3 epsilon | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| CD3E | |
| Cloned human helper T cell line. | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 916 | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| TR66 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes the epsilon-chain of human CD3. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction