missing translation for 'onlineSavingsMsg'
Learn More

CD3 gamma Antibody (CD3G/8794), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-24120MFV500

Product Code. 18588479

  • ONLINE EXCLUSIVE PRICE
    € 736.00
    € 696.15 / 0.10mL
    Save € 39.85
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD3 gamma Monoclonal antibody specifically detects CD3 gamma in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD3 gamma
Monoclonal
mFluor Violet 500 SE
CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ79544, FLJ94613, MGC138597, T3G, T-cell antigen receptor complex, gamma subunit of T3, T-cell receptor T3 gamma chain, T-cell surface glycoprotein CD3 gamma chain
Recombinant fragment (around aa1-182) of human CD3 gamma protein (exact sequence is proprietary)
0.1 mL
Apoptosis, Innate Immunity
917
Store at 4°C in the dark.
IgG2c κ
Immunohistochemistry (Paraffin)
CD3G/8794
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.