missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 gamma Antibody (CD3G/8794), Novus Biologicals™
Mouse Monoclonal Antibody
€ 245.00 - € 486.00
Specifications
| Antigen | CD3 gamma |
|---|---|
| Clone | CD3G/8794 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614104
|
Novus Biologicals
NBP3-23689-20ug |
20 μg |
€ 245.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624964
|
Novus Biologicals
NBP3-23689-100ug |
100 μg |
€ 486.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 gamma Monoclonal antibody specifically detects CD3 gamma in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD3 gamma | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| 917 | |
| IgG2c κ | |
| Protein A or G purified |
| CD3G/8794 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ79544, FLJ94613, MGC138597, T3G, T-cell antigen receptor complex, gamma subunit of T3, T-cell receptor T3 gamma chain, T-cell surface glycoprotein CD3 gamma chain | |
| Recombinant fragment (around aa1-182) of human CD3 gamma protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title