missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human, Clone: UMAB54, liquid, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM500048
This item is not returnable.
View return policy
Description
CD3 epsilon is part of the CD3 subunit complex which is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. The genes encoding the CD3 epsilon, gamma and delta polypeptides are located on chromosome 11. Defects in this gene are associated with T cell immunodeficiency and has been linked to Type 1 Diabetes deficiency in women.Specifications
| CD3 | |
| Monoclonal | |
| Unconjugated | |
| CD3E | |
| IMD18; T3E; TCRE; HCD3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 916 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| UMAB54 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07766 | |
| CD3E | |
| Full length human recombit protein of human CD3E produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction