missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, anti-Human, CoraLite Plus 594, Clone: Hit3a, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-65112-100TESTS
This item is not returnable.
View return policy
Description
The CD3 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD3. This antibody recognizes human antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 594.
Specifications
| CD3 | |
| Hit3a | |
| CoraLite Plus 594 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2a κ |
| Flow Cytometry | |
| 5 μL/Test | |
| CD3E, T3E, TCRE | |
| 100 Tests | |
| 916 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction