Learn More
CD3 Mouse, PE-Cy7, Clone: SK7, enQuireBio™
Product Code. 15941958
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
500 Tests
Unit Size:
100 tests
25 tests
500 tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15941958 25 Tests 25 tests
15931958 100 Tests 100 tests
15951958 500 Tests 500 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15941958 Supplier EnquireBio Supplier No. QAB4PE725Tests
Mouse Monoclonal Antibody
Specifications
Antigen CD3
Applications Flow Cytometry
Classification Monoclonal
Clone SK7
Concentration 5μL/Test
Conjugate PE-Cyanine7
Formulation 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Gene Accession No. NP_000724
Gene Symbols CD3E
Host Species Mouse
Purification Method Protein A/G Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 916
Content And Storage Store at 2-4°C.
Isotype IgG1 κ