Learn More
CD3 Rat anti-Mouse, CoraLite 647, Clone: 17A2, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL647-65077-100TESTS
Description
The CD3 antibody from Proteintech is a rat monoclonal antibody generated with γδtcr-positive t-t hybridoma d1 of mouse CD3. This antibody recognizes mouse antigen. The CD3 antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.
Specifications
| CD3 | |
| Monoclonal | |
| 5 μL/Test | |
| Rat | |
| Primary | |
| Mouse |
| Immunofluorescence, Flow Cytometry | |
| 17A2 | |
| CoraLite 647 | |
| 100 Tests | |
| 12501 | |
| IgG2b |