Learn More
CD3 Rat anti-Mouse, CoraLite 647, Clone: 17A2, Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech CL647-65077-100TESTS
Product Code. 16811116
  • € 120.00 / 100 tests
Description
The CD3 antibody from Proteintech is a rat monoclonal antibody generated with γδtcr-positive t-t hybridoma d1 of mouse CD3. This antibody recognizes mouse antigen. The CD3 antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.
Specifications
Specifications
CD3
Monoclonal
5 μL/Test
Rat
Primary
Mouse
Immunofluorescence, Flow Cytometry
17A2
CoraLite 647
100 Tests
12501
IgG2b