Learn More
CD3 Rat anti-Mouse, FITC, Clone: 17A2, Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech FITC-65077-100UG
Product Code. 16891186
  • € 37.00 / 100µg
Description
The CD3 antibody from Proteintech is a rat monoclonal antibody generated with γδtcr-positive t-t hybridoma d1 of mouse CD3. This antibody recognizes mouse antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
Specifications
CD3
Monoclonal
0.5 mg/mL
Rat
Primary
Mouse
Flow Cytometry
17A2
FITC
100 μg
12501
IgG2b