Learn More
CD3 Rat anti-Mouse, FITC, Clone: 17A2, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech FITC-65077-100UG
Description
The CD3 antibody from Proteintech is a rat monoclonal antibody generated with γδtcr-positive t-t hybridoma d1 of mouse CD3. This antibody recognizes mouse antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
| CD3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| 17A2 | |
| FITC | |
| 100 μg | |
| 12501 | |
| IgG2b |