missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Rat, Qfluor 710, Clone: 17A2, enQuireBio™
Rat Monoclonal Antibody
€ 124.00 - € 263.00
Specifications
Antigen | CD3 |
---|---|
Clone | 17A2 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941808
|
EnquireBio
QAB3-QF710-25UG |
25 μg |
€ 124.00
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931808
|
EnquireBio
QAB3-QF710-100UG |
100 μg |
€ 263.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD3 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
12502 | |
IgG2b κ | |
Protein A/G Purified |
17A2 | |
Flow Cytometry | |
Qfluor 710 | |
RUO | |
NP_033980 | |
Cd3g | |
Primary | |
Store at 2-4°C. |