missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 zeta, p Tyr111 Antibody (EM-55), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37709AF750
This item is not returnable.
View return policy
Description
CD3 zeta Monoclonal specifically detects CD3 zeta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).
Specifications
| CD3 zeta | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD247 | |
| BP1 (pTyr111/123) - KLH. | |
| Primary | |
| Antibody recognizes phosphorylated tyrosine 111 of CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| EM-55 | |
| Western Blot, Flow Cytometry, Flow (Intracellular) | |
| CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA | |
| Mouse | |
| 0.1 mL | |
| 919 | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction