missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 zeta, p Tyr142 Antibody (EM-54), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37708F
This item is not returnable.
View return policy
Description
CD3 zeta Monoclonal specifically detects CD3 zeta in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| CD3 zeta (p Tyr142) | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CD247 | |
| CP1 (pTyr142) - KLH. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 919 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| EM-54 | |
| Flow Cytometry, Flow (Intracellular) | |
| CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Antibody recognizes phosphorylated tyrosine 142 of CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction