Learn More
CD30 Ligand/TNFSF8 Antibody (116614) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1028E
Code produit. 18502835
  • ONLINE EXCLUSIVE PRICE
    € 706.00
    € 667.80 / 0.10mL
    Save € 38.20
    5% Off
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
CD30 Ligand/TNFSF8 Monoclonal antibody specifically detects CD30 Ligand/TNFSF8 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF
TRUSTED_SUSTAINABILITY
Spécification
Spécification
CD30 Ligand/TNFSF8
Monoclonal
DyLight 405
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, Neutralization, CyTOF
116614
Flow Cytometry, Neutralization, CyTOF-ready
CD153, CD153 antigen, CD30 antigen ligand, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8
Mouse myeloma cell line NS0-derived recombinant human CD30 Ligand/TNFSF8, Gln63-Asp234, Accession # P32971
0.1 mL
Apoptosis, Cancer
944
Store at 4°C in the dark.
IgG2b