missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30 Ligand/TNFSF8 Antibody (116621) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10281JF669
This item is not returnable.
View return policy
Description
CD30 Ligand/TNFSF8 Monoclonal antibody specifically detects CD30 Ligand/TNFSF8 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF
Specifications
| CD30 Ligand/TNFSF8 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD153, CD153 antigen, CD30 antigen ligand, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8 | |
| Mouse myeloma cell line NS0-derived human CD30 Ligand/TNFSF8., Gln63-Asp234, Accession # P32971 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 116621 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 944 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction