missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300a/LMIR1 Antibody (232612), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2640L
This item is not returnable.
View return policy
Description
CD300a/LMIR1 Monoclonal antibody specifically detects CD300a/LMIR1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| CD300a/LMIR1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 232612 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD300a antigenCMRF35H9, CD300a molecule, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, Irp60, leukocyte membrane antigen | |
| Y3 rat myeloid cell line transfected with human CD300a/LMIR1 | |
| 0.1 mL | |
| Mast Cell Markers | |
| 11314 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction