missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300c Antibody (03), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06451B
This item is not returnable.
View return policy
Description
CD300c Monoclonal antibody specifically detects CD300c in Human samples. It is validated for Flow Cytometry
Specifications
| CD300c | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 03 | |
| Flow Cytometry | |
| CD300c antigenCMRF35 antigen, CD300c molecule, CLM-6, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IgSF16, IGSF16CD300 antigen-like family member C, Immunoglobulin superfamily member 16, LIR | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD300c (NP_006669.1; Met1-Arg183). | |
| 0.1 mL | |
| Immunology | |
| 10871 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction