missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300e/LMIR6 Antibody (UP-H2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62230DL594
This item is not returnable.
View return policy
Description
CD300e/LMIR6 Monoclonal antibody specifically detects CD300e/LMIR6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay
Specifications
| CD300e/LMIR6 | |
| Monoclonal | |
| DyLight 594 | |
| CD300 antigen like family member E, CD300 antigen-like family member E, CD300e antigenPIgR-2, CD300e molecule, CD300LEPIgR2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM2IREM-2, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2 | |
| CD300e-HA-transfected cells | |
| 0.1 mL | |
| Immunology | |
| 342510 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay | |
| UP-H2 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction