missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300f/LMIR3/CD300LF Antibody (234903), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2774F
This item is not returnable.
View return policy
Description
CD300f/LMIR3/CD300LF Monoclonal antibody specifically detects CD300f/LMIR3/CD300LF in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| CD300f/LMIR3/CD300LF | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 146722 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 234903 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CLM1, CLM-1, IgSF13, IREM1, IREM-1, NKIR | |
| Y3 rat myeloid cell line transfected with human CD300f/LMIR3 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction