missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300LG/Nepmucin Antibody (102), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90215CL1
This item is not returnable.
View return policy
Description
CD300LG/Nepmucin Monoclonal antibody specifically detects CD300LG/Nepmucin in Human samples. It is validated for ELISA
Specifications
| CD300LG/Nepmucin | |
| Monoclonal | |
| CoraFluor 1 | |
| CD300 antigen like family member G, CD300 antigen-like family member G, CD300 molecule like family member g, CD300 molecule-like family member g, CD300g antigen, CLM-9, CLM9Trem4, CMRF35-like molecule 9, NEPMUCIN, TREM4, TREM-4, Triggering receptor expressed on myeloid cells 4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CD300LG/Nepmucin (Accession#: AAH25395.1; Met1-Arg247). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 102 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 146894 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction