Learn More
CD305 (LAIR1) Monoclonal Antibody (113), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
€ 10000.00 - € 14910.00
Specifications
| Antigen | CD305 (LAIR1) |
|---|---|
| Clone | 113 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17171202
|
Affymetrix eBioscience
16-3051-025 |
25 mg |
€ 10000.00
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17181202
|
Affymetrix eBioscience
16-3051-050 |
50 mg | |||||||
Description
CD305 or Leukocyte-associated Ig-like receptor-1 (LAIR-1) is a 46 kD cell surface molecule that functions as an inhibitory collagen-receptor on the majority of cells of the immune system. The mouse LAIR-1 molecule is expressed primarily on monocytes/macrophages and also on T cells, NK cells and dendritic cells. Unlike human B cells , mouse B cells do not express LAIR-1. The cytoplasmic tail of mouse LAIR-1 can be phosphorylated leading to the recruitment of SH2-containing tyrosine phosphatase(SHP)-2 and C-terminal Src kinase. Applications Reported: This 113 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB). It has also been reported for use in functional studies. Applications Tested: This 113 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD305 (LAIR1) Monoclonal antibody specifically detects CD305 (LAIR1) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation, Western BlotSpecifications
| CD305 (LAIR1) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; LAIR-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 113 | |
| Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot | |
| Functional Grade | |
| Armenian Hamster | |
| Mouse | |
| Q8BG84 | |
| 52855 | |
| Primary | |
| 4° C | |
| LAIR1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.