missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31/PECAM-1 Antibody (rPECAM1/8832), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23736-100ug
This item is not returnable.
View return policy
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD31/PECAM-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Recombinant fragment (around aa 625-738) of human CD31/PECAM-1 protein (exact sequence is proprietary) | |
| 100 μg | |
| Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
| 5175 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rPECAM1/8832 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction